3-Bromobenzo[b]thiophene 95%

Supplier: Thermo Scientific Chemicals
Warning

H32948-09 H32948-18
AAH32948-09EA 92.5 USD
AAH32948-09 AAH32948-18
3-Bromobenzo[b]thiophene 95%
3-Bromobenzo[b]thiophene

Purity: 95% CAS No.: 7342-82-7 Molecular Formula: C8H5BrS Formula Weight: 213.10 Refractive Index: 1.6680 Boiling Point: 72-74°/0.27mm MDL No.: MFCD00023009

Formula: C₈H₅BrS
MW: 213.10 g/mol
Boiling Pt: 72…74 °C/0.27 mm
Density: 1.629
Storage Temperature: Ambient
MDL Number: MFCD00023009
CAS Number: 7342-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR