2-Methyl-1,3-benzothiazol-5-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

H32937-03 H32937-09
AAH32937-03EA 38 USD
AAH32937-03 AAH32937-09
2-Methyl-1,3-benzothiazol-5-ol 97%
2-Methyl-1,3-benzothiazol-5-ol

Purity: 97% CAS No.: 68867-14-1 Molecular Formula: C8H7NOS Formula Weight: 165.21 Melting Point: 185-187° MDL No.: MFCD00192276

Formula: C₈H₇NOS
MW: 165.22 g/mol
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00192276
CAS Number: 68867-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR