1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1,3,5-Trimethylpyrazole-4-boronic acid Pinacol ester, 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1,3,5-Trimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H32930-03 H32930-MD
AAH32930-03EA 198 USD
AAH32930-03 AAH32930-MD
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester

Purity: 95% CAS No.: 844891-04-9 Molecular Formula: C12H21BN2O2 Formula Weight: 236.12 MDL No.: MFCD06659062

Formula: C₁₂H₂₁BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06659062
CAS Number: 844891-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR