6-Bromo-m-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H32927-03 H32927-06
AAH32927-03EA 22.9 USD
AAH32927-03 AAH32927-06
6-Bromo-m-toluic acid 98%
6-Bromo-m-toluic acid

Purity: 98% CAS No.: 6967-82-4 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 MDL No.: MFCD00079722

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079722
CAS Number: 6967-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR