Ethyl-5-oxazolecarboxylate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl oxazole-5-carboxylate

H32924-06 H32924-14
AAH32924-06EA 93.4 USD
AAH32924-06 AAH32924-14
Ethyl-5-oxazolecarboxylate 98%
Ethyl 5-oxazolecarboxylate

Purity: 98% CAS No.: 118994-89-1 Molecular Formula: C6H7NO3 Formula Weight: 141.13 Refractive Index: 1.4690 Boiling Point: 60°/2.25mm MDL No.: MFCD04114930

Formula: C₆H₇NO₃
Boiling Pt: 60 °C/2.25 mm
Storage Temperature: Ambient
MDL Number: MFCD04114930
CAS Number: 118994-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR