5-Chloro-2-methylbenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Chloro-2-methylbenzene-1-sulphonyl chloride, 4-Chlorotoluene-2-sulphonyl chloride

H32912-03 H32912-06
AAH32912-03EA 92 USD
AAH32912-03 AAH32912-06
5-Chloro-2-methylbenzenesulfonyl chloride 97%
5-Chloro-2-methylbenzenesulphonyl chloride

Purity: 97% CAS No.: 34981-38-9 Molecular Formula: C7H6Cl2O2S Formula Weight: 225.09 Refractive Index: 1.5697

Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09742738
CAS Number: 34981-38-9
EINECS: 252-306-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR