3-Chloro-1-fluoro-4-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

H32906-03 H32906-06 H32906-14
AAH32906-03EA 43.3 USD
AAH32906-03 AAH32906-06 AAH32906-14
3-Chloro-1-fluoro-4-nitrobenzene 98%
3-Chloro-1-fluoro-4-nitrobenzene

Purity: 98% CAS No.: 2106-50-5 Molecular Formula: C6H3ClFNO2 Formula Weight: 175.55 Physical Form: Solid Appearance: Yellow Odor: Characteristic Melting Point: 33-37° UN No.: 2811 MDL No.: MFCD03412200

Formula: C₆H₃ClFNO₂
Melting Pt: 33…37 °C
Storage Temperature: Ambient
MDL Number: MFCD03412200
CAS Number: 2106-50-5
EINECS: 218-286-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR