Ethyl-1,3-benzothiazole-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzothiazole-2-carboxylic acid ethyl ester, Ethyl 1,3-benzothiazole-2-carboxylate, Ethyl Benzothiazole-2-carboxylate

H32904-06 H32904-14
AAH32904-06EA 66.7 USD
AAH32904-06 AAH32904-14
Ethyl-1,3-benzothiazole-2-carboxylate 98%
Ethyl-1,3-benzothiazole-2-carboxylate

Purity: 98% CAS No.: 32137-76-1 Molecular Formula: C10H9NO2S Formula Weight: 207.25 Melting Point: 68-72° MDL No.: MFCD00848360

Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00848360
CAS Number: 32137-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR