Trimethylamine 1 M in THF
Supplier: Thermo Scientific Chemicals
|
Danger
|
H32899-AE
H32899-AP
AAH32899-AEEA
187.73
USD
AAH32899-AE
AAH32899-AP
Trimethylamine 1 M in THF
Trimethylamine
CAS No.: 75-50-3
Molecular Formula: C3H9N
Formula Weight: 59.11
UN No.: 1993
MDL No.: MFCD00008327
Formula:
C₃H₉N Density: 0.850 Storage Temperature: Ambient |
MDL Number:
MFCD00008327 CAS Number: 75-50-3 EINECS: 200-875-0 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...