Methyl-3-(trifluoromethyl)phenylacetate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-(3-(trifluoromethyl)phenyl)acetate

H32883-03 H32883-06
AAH32883-03EA 44.6 USD
AAH32883-03 AAH32883-06 AAH32883-14
Methyl-3-(trifluoromethyl)phenylacetate 95%
Methyl-3-(trifluoromethyl)phenylacetate

Purity: 95% CAS No.: 62451-84-7 Molecular Formula: C10H9F3O2 Formula Weight: 218.18 Refractive Index: 1.4500 Boiling Point: 211° MDL No.: MFCD04039762

Formula: C₁₀H₉F₃O₂
MW: 218.175 g/mol
Boiling Pt: 211 °C
Density: 1.24
Storage Temperature: Ambient
MDL Number: MFCD04039762
CAS Number: 62451-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR