3-(Benzyloxy)benzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Benzyloxybenzoic acid
H32882-03
H32882-06
H32882-14
AAH32882-03EA
42.3
USD
AAH32882-03
AAH32882-06
AAH32882-14
3-(Benzyloxy)benzoic acid 98%
3-(Benzyloxy)benzoic acid
Purity: 98%
CAS No.: 69026-14-8
Molecular Formula: C14H12O3
Formula Weight: 228.25
Melting Point: 133-135°
MDL No.: MFCD00527901
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Melting Pt: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00527901 CAS Number: 69026-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...