2-Hydroxy-3-(trifluoromethyl)benzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Trifluoromethyl)salicylaldehyde

H32858-03 H32858-MD
AAH32858-03EA 321 USD
AAH32858-03 AAH32858-MD
2-Hydroxy-3-(trifluoromethyl)benzaldehyde 98+%
2-Hydroxy-3-(trifluoromethyl)benzaldehyde

Purity: 98+% CAS No.: 336628-67-2 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Melting Point: 54-59° MDL No.: MFCD03788529

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Melting Pt: 54…59 °C
Storage Temperature: Ambient
MDL Number: MFCD03788529
CAS Number: 336628-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR