3-Nitro-5-(trifluoromethyl)aniline 98%

Supplier: Thermo Scientific Chemicals
Danger

H32823-03 H32823-09
AAH32823-03EA 92.2 USD
AAH32823-03 AAH32823-09
3-Nitro-5-(trifluoromethyl)aniline 98%
3-Nitro-5-(trifluoromethyl)aniline

Purity: 98% CAS No.: 401-94-5 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 77-81° UN No.: 2811 MDL No.: MFCD00061229

Formula: C₇H₅F₃N₂O₂
MW: 206.12
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00061229
CAS Number: 401-94-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR