2-Nitro-4-(trifluoromethyl)benzyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Bromomethyl)-2-nitro-4-(trifluoromethyl)benzene
H32802-03
H32802-06
AAH32802-03EA
88.4
USD
AAH32802-03
AAH32802-06
2-Nitro-4-(trifluoromethyl)benzyl bromide 97%
2-Nitro-4-(trifluoromethyl)benzyl bromide
Purity: 97%
CAS No.: 162333-02-0
Molecular Formula: C8H5BrF3NO3
Formula Weight: 284.03
Physical Form: Solid
Appearance: White to brown
Odor: Pungent
Melting Point: 36-39°
UN No.: 3261
MDL No.: MFCD03095413
Formula:
C₈H₅BrF₃NO₂ MW: 284.03 g/mol Melting Pt: 36…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095413 CAS Number: 162333-02-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...