tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-DL-Prolinol, tert-Butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate

H32786-03 H32786-06
AAH32786-03EA 36.6 USD
AAH32786-03 AAH32786-06
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate 98%
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Purity: 98% CAS No.: 170491-63-1 Molecular Formula: C10H19NO3 Formula Weight: 201.27 Melting Point: 52-54° MDL No.: MFCD01456556

Formula: C₁₀H₁₉NO₃
MW: 201.3 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD01456556
CAS Number: 170491-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR