2-Fluoro-6-methoxybenzamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32774-03
H32774-06
H32774-MD
AAH32774-03EA
80.1
USD
AAH32774-03
AAH32774-06
AAH32774-MD
2-Fluoro-6-methoxybenzamide 98%
2-Fluoro-6-methoxybenzamide
Purity: 98%
CAS No.: 529512-81-0
Molecular Formula: C8H8FNO2
Formula Weight: 169.15
Melting Point: 190-194°
MDL No.: MFCD00277474
Formula:
C₈H₈FNO₂ Melting Pt: 190…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00277474 CAS Number: 529512-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...