(2,3-Dichlorophenyl)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Dichlorophenylacetic acid
H32763-06
H32763-14
AAH32763-06EA
27.2
USD
AAH32763-06
AAH32763-14
(2,3-Dichlorophenyl)acetic acid 98%
(2,3-Dichlorophenyl)acetic acid
Purity: 98%
CAS No.: 10236-60-9
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.04
Odor: Odorless
Melting Point: 132-136°
MDL No.: MFCD01861393
Formula:
(C₆H₃Cl₂)CH₂COOH MW: 205.04 g/mol Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861393 CAS Number: 10236-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...