5-Fluoro-2-nitrobenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H32756-03
H32756-06
AAH32756-03EA
66.4
USD
AAH32756-03
AAH32756-06
5-Fluoro-2-nitrobenzonitrile 97%
5-Fluoro-2-nitrobenzonitrile
Purity: 97%
CAS No.: 50594-78-0
Molecular Formula: C7H3FN2O2
Formula Weight: 166.11
Melting Point: 101-105°
UN No.: 3439
MDL No.: MFCD01632196
Formula:
C₇H₃FN₂O₂ Melting Pt: 101…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632196 CAS Number: 50594-78-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...