Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

H32755-06 H32755-14
AAH32755-06EA 73.4 USD
AAH32755-06 AAH32755-14
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 98%
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Purity: 98% CAS No.: 108354-78-5 Molecular Formula: C10H13NO2S Formula Weight: 211.28 Melting Point: 126-129° MDL No.: MFCD00130099

Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00130099
CAS Number: 108354-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR