4,5-Dimethoxy-2-nitrophenylacetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H32747-03
H32747-MD
AAH32747-03EA
96
USD
AAH32747-03
AAH32747-MD
4,5-Dimethoxy-2-nitrophenylacetonitrile 98%
4,5-Dimethoxy-2-nitrophenylacetonitrile
Purity: 98%
CAS No.: 17354-04-0
Molecular Formula: C10H10N2O4
Formula Weight: 222.20
Melting Point: 110-112°
UN No.: 3439
MDL No.: MFCD00033967
Formula:
C₁₀H₁₀N₂O₄ Melting Pt: 110…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00033967 CAS Number: 17354-04-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...