3-Methylbenzoic anhydride 97%

Supplier: Thermo Scientific Chemicals
Warning

H32739-03 H32739-06
AAH32739-03EA 34.5 USD
AAH32739-03 AAH32739-06
3-Methylbenzoic anhydride 97%
3-Methylbenzoic anhydride

Purity: 97% CAS No.: 21436-44-2 Molecular Formula: C16H14O3 Formula Weight: 254.29 Melting Point: 66-70° MDL No.: MFCD00048083

Formula: C₁₆H₁₄O₃
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00048083
CAS Number: 21436-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR