Methyl-4-(hydroxymethyl)picolinate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, Methyl 4-(hydroxymethyl)picolinate
H32731-03
H32731-06
AAH32731-03EA
193
USD
AAH32731-03
AAH32731-06
AAH32731-MD
Methyl-4-(hydroxymethyl)picolinate 95%
Methyl 4-(hydroxymethyl)picolinate
Purity: 95%
CAS No.: 317335-15-2
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Melting Point: 74-76°
MDL No.: MFCD09834350
Formula:
C₈H₉NO₃ MW: 167.16 g/mol Melting Pt: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD09834350 CAS Number: 317335-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...