tert-Butyl-2-((diphenylmethylene)amino)acetate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate
H32709-03
H32709-06
AAH32709-03EA
87.7
USD
AAH32709-03
AAH32709-06
tert-Butyl-2-((diphenylmethylene)amino)acetate 99%
tert-Butyl-2-((diphenylmethylene)amino)acetate
Purity: 99%
CAS No.: 81477-94-3
Molecular Formula: C19H21NO2
Formula Weight: 295.38
Melting Point: 113-115°
MDL No.: MFCD00134280
Formula:
C₁₉H₂₁NO₂ Melting Pt: 113…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134280 CAS Number: 81477-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...