4,5-Dimethyl-o-phenylenediamine, (∼15% H₂O) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4,5-Dimethylbenzene-1,2-diamine, 4,5-Dimethyl-1,2-phenylenediamine, 4,5-Diamino-1,2-dimethylbenzene, 1,2-Diamino-4,5-dimethylbenzene
H32695-06
H32695-14
AAH32695-06EA
68.6
USD
AAH32695-06
AAH32695-14
4,5-Dimethyl-o-phenylenediamine, (∼15% H₂O) 98%
4,5-Dimethyl-o-phenylenediamine
Purity: 98%
CAS No.: 3171-45-7
Molecular Formula: C8H12N2
Formula Weight: 136.20
Storage Temperature: 2°C to 8°C
Melting Point: 127-129°
UN No.: 2811
MDL No.: MFCD00007729
Formula:
C₈H₁₂N₂ Melting Pt: 127…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007729 CAS Number: 3171-45-7 EINECS: 221-635-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...