[1,2,4]Triazolo[4,3-a]pyridine-3-thiol 96%

Supplier: Thermo Scientific Chemicals
Warning

H32694-03 H32694-06
AAH32694-03EA 44.7 USD
AAH32694-03 AAH32694-06
[1,2,4]Triazolo[4,3-a]pyridine-3-thiol 96%
[1,2,4]Triazolo[4,3-a]pyridine-3-thiol

Purity: 96% CAS No.: 6952-68-7 Molecular Formula: C6H5N3S Formula Weight: 151.19 Melting Point: 209-210° MDL No.: MFCD03783345

Formula: C₆H₅N₃S
Melting Pt: 209…210 °C
Storage Temperature: Ambient
MDL Number: MFCD03783345
CAS Number: 6952-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR