4,5-Difluorosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-4,5-difluorobenzaldehyde, 4,5-Difluoro-2-hydroxybenzaldehyde

H32680-03 H32680-MD
AAH32680-03EA 191 USD
AAH32680-03 AAH32680-MD
4,5-Difluorosalicylaldehyde 98%
4,5-Difluorosalicylaldehyde

Purity: 98% CAS No.: 199287-52-0 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Melting Point: 68-72° MDL No.: MFCD08458005

Formula: C₇H₄F₂O₂
MW: 158.11 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD08458005
CAS Number: 199287-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR