3,5-Dinitrobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

H32669-06 H32669-14 H32669-22
AAH32669-06EA 87.9 USD
AAH32669-06 AAH32669-14 AAH32669-22
3,5-Dinitrobenzyl alcohol 98%
3,5-Dinitrobenzyl alcohol

Purity: 98% CAS No.: 71022-43-0 Molecular Formula: C7H6N2O5 Formula Weight: 198.13 Melting Point: 88-91° Beilstein Reference: 2054388

Formula: C₇H₆N₂O₅
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007235
CAS Number: 71022-43-0
EINECS: 275-131-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR