1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-1-fluoro-4-(trifluoromethoxy)benzene
H32650-03
H32650-06
H32650-14
AAH32650-03EA
21.8
USD
AAH32650-03
AAH32650-06
AAH32650-14
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene 98%
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene
Purity: 98%
CAS No.: 286932-57-8
Molecular Formula: C7H3BrF4O
Formula Weight: 259.00
Physical Form: Liquid
Appearance: Colourless to yellow
MDL No.: MFCD02181186
Formula:
C₇H₃BrF₄O MW: 259.00 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02181186 CAS Number: 286932-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...