4-Bromo-2-(trifluoromethoxy)anisole 98%

Supplier: Thermo Scientific Chemicals
Warning

H32645-03 H32645-06
AAH32645-03EA 41.4 USD
AAH32645-03 AAH32645-06
4-Bromo-2-(trifluoromethoxy)anisole 98%
4-Bromo-2-(trifluoromethoxy)anisole

Purity: 98% CAS No.: 853771-88-7 Molecular Formula: C8H6BrF3O2 Formula Weight: 271.04 MDL No.: MFCD09907950

Formula: C₈H₆BrF₃O₂
MW: 271.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907950
CAS Number: 853771-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR