3-Bromo-5-methoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H32635-03 H32635-06
AAH32635-03EA 38 USD
AAH32635-03 AAH32635-06
3-Bromo-5-methoxybenzoic acid 98%
3-Bromo-5-methoxybenzoic acid

Purity: 98% CAS No.: 157893-14-6 Molecular Formula: C8H7BrO3 Formula Weight: 231.05 MDL No.: MFCD00266858

Formula: C₈H₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00266858
CAS Number: 157893-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR