4-Amino-α,α,α-trifluoro-o-toluic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-2-(trifluoromethyl)benzoic acid

H32626-03 H32626-06
AAH32626-03EA 19.5 USD
AAH32626-03 AAH32626-06
4-Amino-α,α,α-trifluoro-o-toluic acid 97+%
4-Amino-α,α,α-trifluoro-o-toluic acid

Purity: 97+% CAS No.: 393-06-6 Molecular Formula: C8H6F3NO2 Formula Weight: 205.14 Melting Point: 183-186° MDL No.: MFCD03407959

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD03407959
CAS Number: 393-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR