2-Fluoro-4-(trifluoromethyl)nicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid

H32618-03 H32618-MD
AAH32618-03EA 115 USD
AAH32618-03 AAH32618-MD
2-Fluoro-4-(trifluoromethyl)nicotinic acid 97%
2-Fluoro-4-(trifluoromethyl)nicotinic acid

Purity: 97% Molecular Formula: C7H3F4NO2 Formula Weight: 209.10 Melting Point: 91-93° MDL No.: MFCD10687403

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD10687403
CAS Number: 1040681-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR