2,4-Difluoro-5-nitrobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32617-03
H32617-06
H32617-14
AAH32617-03EA
37.6
USD
AAH32617-03
AAH32617-06
AAH32617-14
2,4-Difluoro-5-nitrobenzoic acid 97%
2,4-Difluoro-5-nitrobenzoic acid
Purity: 97%
CAS No.: 153775-33-8
Molecular Formula: C7H3F2NO4
Formula Weight: 203.10
Melting Point: 125-133°
MDL No.: MFCD04115713
Formula:
C₇H₃F₂NO₄ Melting Pt: 125…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115713 CAS Number: 153775-33-8 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...