Ammonium O,O'-dimethyldithiophosphate 95%

Supplier: Thermo Scientific Chemicals
Warning

H32592-03 H32592-09
AAH32592-03EA 48.6 USD
AAH32592-03 AAH32592-09
Ammonium O,O'-dimethyldithiophosphate 95%
Ammonium O,O'-dimethyldithiophosphate

Purity: 95% CAS No.: 1066-97-3 Molecular Formula: C2H10NO2PS2 Formula Weight: 175.21 Odor: Weak Melting Point: 146-150° MDL No.: MFCD09753116

Formula: C₂H₁₀NO₂PS₂
MW: 175.21 g/mol
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD09753116
CAS Number: 1066-97-3
EINECS: 213-922-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR