Bis(4-fluorophenyl) disulfide 98%

Supplier: Thermo Scientific Chemicals
Warning

H32589-03 H32589-06
AAH32589-03EA 70.7 USD
AAH32589-03 AAH32589-06
Bis(4-fluorophenyl) disulfide 98%
Bis(4-fluorophenyl)disulphide

Purity: 98% CAS No.: 405-31-2 Molecular Formula: C12H8F2S2 Formula Weight: 254.31 Boiling Point: 141°/1mm MDL No.: MFCD00816758

Formula: C₁₂H₈F₂S₂
MW: 254.32 g/mol
Boiling Pt: 141 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD00816758
CAS Number: 405-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR