5-Methoxy-2-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H32586-06 H32586-03
AAH32586-06EA 231 USD
AAH32586-06 AAH32586-03
5-Methoxy-2-(trifluoromethyl)pyridine 97%
5-Methoxy-2-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 216766-13-1 Molecular Formula: C7H6F3NO Formula Weight: 177.13 MDL No.: MFCD09864699

Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864699
CAS Number: 216766-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR