2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H32580-03 H32580-09
AAH32580-03EA 37.2 USD
AAH32580-03 AAH32580-09
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene 97%
2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene

Purity: 97% CAS No.: 118754-55-5 Molecular Formula: C7H2BrCl2F3O Formula Weight: 309.90 Physical Form: Liquid Appearance: Colorless tp pale yellow Odor: Characteristic MDL No.: MFCD03412210

Formula: C₇H₂BrCl₂F₃O
MW: 309.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412210
CAS Number: 118754-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR