N-Boc-ethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl N-ethylcarbamate, tert-Butyl ethylcarbamate

H32575-03 H32575-06
AAH32575-03EA 69.8 USD
AAH32575-03 AAH32575-06
N-Boc-ethylamine 97%
N-Boc-ethylamine

Purity: 97% CAS No.: 38267-76-4 Molecular Formula: C7H15NO2 Formula Weight: 145.20 Melting Point: 38-41° MDL No.: MFCD12923020

Formula: C₇H₁₅NO₂
MW: 145.20 g/mol
Melting Pt: 38…41 °C
Storage Temperature: Ambient
MDL Number: MFCD12923020
CAS Number: 38267-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR