Ethyl-2-bromo-4,4,4-trifluorobutyrate 97%

Supplier: Thermo Scientific Chemicals
Danger

H32572-03 H32572-MD
AAH32572-03EA 179 USD
AAH32572-03 AAH32572-MD
Ethyl-2-bromo-4,4,4-trifluorobutyrate 97%
Ethyl-2-bromo-4,4,4-trifluorobutyrate

Purity: 97% CAS No.: 367-33-9 Molecular Formula: C6H8BrF3O2 Formula Weight: 249.03 Boiling Point: 67°/18mm UN No.: 3265 MDL No.: MFCD10687235

Formula: C₆H₈BrF₃O₂
MW: 249.03 g/mol
Boiling Pt: 67 °C/18 mm
Storage Temperature: Ambient
MDL Number: MFCD10687235
CAS Number: 367-33-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR