5-Bromo-2-isopropoxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32564-03
H32564-06
AAH32564-03EA
105
USD
AAH32564-03
AAH32564-06
5-Bromo-2-isopropoxypyridine 97%
5-Bromo-2-isopropoxypyridine
Purity: 97%
CAS No.: 870521-31-6
Molecular Formula: C8H10BrNO
Formula Weight: 216.08
Refractive Index: 1.5305
Formula:
C₈H₁₀BrNO Storage Temperature: Ambient |
MDL Number:
MFCD07368869 CAS Number: 870521-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...