3-Phenyl-1H-pyrazole-4-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32560-03
H32560-06
H32560-MD
AAH32560-03EA
128
USD
AAH32560-03
AAH32560-06
AAH32560-MD
3-Phenyl-1H-pyrazole-4-carboxylic acid 97%
3-Phenyl-1H-pyrazole-4-carboxylic acid
Purity: 97%
CAS No.: 5504-65-4
Molecular Formula: C10H8N2O2
Formula Weight: 188.19
Melting Point: 280-285°
MDL No.: MFCD06798067
Formula:
C₁₀H₈N₂O₂ Melting Pt: 280…285 °C Storage Temperature: Ambient |
MDL Number:
MFCD03834501 CAS Number: 5504-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...