3-Methyl-1H-pyrazole-4-carbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Formyl-3-methylpyrazole, 3-Methylpyrazole-4-carboxaldehyde
H32547-03
H32547-MD
AAH32547-03EA
99.4
USD
AAH32547-03
AAH32547-MD
3-Methyl-1H-pyrazole-4-carbaldehyde 97%
3-Methyl-1H-pyrazole-4-carbaldehyde
Purity: 97%
CAS No.: 112758-40-4
Molecular Formula: C5H6N2O
Formula Weight: 110.12
MDL No.: MFCD01313822
Formula:
C5H6N2O MW: 110.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01313822 CAS Number: 112758-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...