2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32544-03
H32544-09
AAH32544-03EA
36.94
USD
AAH32544-03
AAH32544-09
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 97%
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Purity: 97%
CAS No.: 4651-91-6
Molecular Formula: C9H10N2S
Formula Weight: 178.26
Melting Point: 141-144°
MDL No.: MFCD00128278
Formula:
C₉H₁₀N₂S Melting Pt: 141…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128278 CAS Number: 4651-91-6 EINECS: 225-085-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...