2,5-Dimethylthiophene-3-boronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2,5-Dimethylthiophen-3-yl)boronic acid

H32543-03 H32543-MD
AAH32543-03EA 268 USD
AAH32543-03 AAH32543-MD
2,5-Dimethylthiophene-3-boronic acid 95%
2,5-Dimethylthiophene-3-boronic acid

Purity: 95% CAS No.: 162607-23-0 Molecular Formula: C6H9BO2S Formula Weight: 156.01 Storage Temperature: 2°C to 8°C MDL No.: MFCD09837620

Formula: C₆H₉BO₂S
MW: 156.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09837620
CAS Number: 162607-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR