Ethyl-2-amino-4-oxazolecarboxylate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 2-aminooxazole-4-carboxylate
H32542-03
H32542-06
AAH32542-03EA
54.2
USD
AAH32542-03
AAH32542-06
Ethyl-2-amino-4-oxazolecarboxylate 95%
Ethyl 2-amino-4-oxazolecarboxylate
Purity: 95%
CAS No.: 177760-52-0
Molecular Formula: C6H8N2O3
Formula Weight: 156.14
Melting Point: 124-128°
MDL No.: MFCD06659907
Formula:
C₆H₈N₂O₃ MW: 156.14 g/mol Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659907 CAS Number: 177760-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...