5-Chloro-2-hydroxybenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Chloro-2-(hydroxymethyl)phenol
H32533-03
H32533-09
AAH32533-03EA
41.7
USD
AAH32533-03
AAH32533-09
5-Chloro-2-hydroxybenzyl alcohol 98%
5-Chloro-2-hydroxybenzyl alcohol
Purity: 98%
CAS No.: 5330-38-1
Molecular Formula: C7H7ClO2
Formula Weight: 158.58
Melting Point: 84-87°
MDL No.: MFCD00021967
Formula:
C₇H₇ClO₂ MW: 158.59 g/mol Melting Pt: 84…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021967 CAS Number: 5330-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...