(+/-)-2-(Trifluoromethyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Trifluoromethyl)piperazine

H32524-03 H32524-MD
AAH32524-03EA 188 USD
AAH32524-03 AAH32524-MD
(+/-)-2-(Trifluoromethyl)piperazine 97%
(+/-)-2-(Trifluoromethyl)piperazine

Purity: 97% CAS No.: 131922-05-9 Molecular Formula: C5H9F3N2 Formula Weight: 154.13 Melting Point: 94-97° UN No.: 3259 MDL No.: MFCD07373382

Formula: C₅H₉F₃N₂
MW: 154.13 g/mol
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD07373382
CAS Number: 131922-05-9
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR