4-Bromo-3-fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

H32488-06 H32488-14
AAH32488-06EA 61.9 USD
AAH32488-06 AAH32488-14
4-Bromo-3-fluorophenol 98%
4-Bromo-3-fluorophenol

Purity: 98% CAS No.: 121219-03-2 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Melting Point: 71-73° MDL No.: MFCD00051907

Formula: C₆H₄BrFO
MW: 191
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00051907
CAS Number: 121219-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR