3-Acetylbenzenesulphonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Acetylbenzene-1-sulphonyl chloride
H32447-03
H32447-06
AAH32447-03EA
96.9
USD
AAH32447-03
AAH32447-06
3-Acetylbenzenesulphonyl chloride 97%
3-Acetylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 73035-16-2
Molecular Formula: C8H7ClO3S
Formula Weight: 218.66
Melting Point: 42-43°
UN No.: 3261
MDL No.: MFCD03424979
Formula:
C₈H₇ClO₃S Melting Pt: 42…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD03424979 CAS Number: 73035-16-2 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...