6-Methoxy-2-methylbenzothiazole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Methoxy-2-methylbenzo[d]thiazole

H32425-03 H32425-06
AAH32425-03EA 88.8 USD
AAH32425-03 AAH32425-06
6-Methoxy-2-methylbenzothiazole 97%
6-Methoxy-2-methylbenzothiazole

Purity: 97% CAS No.: 2941-72-2 Molecular Formula: C9H9NOS Formula Weight: 179.24 Flash Point: 109°(228°F)

Formula: C₉H₉NOS
Density: 1.204
Flash Pt: 109 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005795
CAS Number: 2941-72-2
EINECS: 220-931-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR